Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGCCTTTGGAGGTCGCACCACA[A/G]CACCGGGGCCGCTTGCTGGCTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 602930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139025.4 | 416 | Intron | NP_620594.1 | |||
NM_139026.4 | 416 | Intron | NP_620595.1 | |||
NM_139027.4 | 416 | Intron | NP_620596.2 | |||
XM_011518176.2 | 416 | Intron | XP_011516478.1 | |||
XM_011518178.1 | 416 | Intron | XP_011516480.1 | |||
XM_011518179.1 | 416 | Intron | XP_011516481.1 | |||
XM_017014232.1 | 416 | Intron | XP_016869721.1 | |||
XM_017014233.1 | 416 | Intron | XP_016869722.1 | |||
XM_017014234.1 | 416 | Intron | XP_016869723.1 | |||
XM_017014235.1 | 416 | Intron | XP_016869724.1 |
REXO4 - REX4 homolog, 3'-5' exonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279349.1 | 416 | Missense Mutation | GCT,GTT | A,V 55 | NP_001266278.1 | |
NM_001279350.1 | 416 | UTR 5 | NP_001266279.1 | |||
NM_001279351.1 | 416 | Intron | NP_001266280.1 | |||
NM_020385.3 | 416 | Missense Mutation | GCT,GTT | A,V 55 | NP_065118.2 |