Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTCCCGCATGGTCTTGAGGTC[A/G]AACACCAGCAGGTCAATCTCCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120930 MIM: 609072 MIM: 612905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8G - complement component 8, gamma polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXW5 - F-box and WD repeat domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018998.3 | 1524 | Silent Mutation | TTC,TTT | F,F 448 | NP_061871.1 | |
XM_005266089.2 | 1524 | Silent Mutation | TTC,TTT | F,F 458 | XP_005266146.2 | |
XM_011518789.1 | 1524 | Silent Mutation | TTC,TTT | F,F 440 | XP_011517091.1 | |
XM_017014812.1 | 1524 | Silent Mutation | TTC,TTT | F,F 430 | XP_016870301.1 |
LCN12 - lipocalin 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |