Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGCTGGCAGCACCCAGCAGGG[C/T]GGGGTCAACCAAATCCGAAACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118960 MIM: 603824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLTA - clathrin light chain A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNE - glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128227.2 | 2179 | Missense Mutation | ACC,GCC | T,A 736 | NP_001121699.1 | |
NM_001190383.1 | 2179 | Missense Mutation | ACC,GCC | T,A 631 | NP_001177312.1 | |
NM_001190384.1 | 2179 | Missense Mutation | ACC,GCC | T,A 595 | NP_001177313.1 | |
NM_001190388.1 | 2179 | Missense Mutation | ACC,GCC | T,A 700 | NP_001177317.1 | |
NM_005476.5 | 2179 | Missense Mutation | ACC,GCC | T,A 705 | NP_005467.1 | |
XM_005251334.4 | 2179 | Missense Mutation | ACC,GCC | T,A 685 | XP_005251391.1 | |
XM_017014167.1 | 2179 | Missense Mutation | ACC,GCC | T,A 705 | XP_016869656.1 | |
XM_017014168.1 | 2179 | Missense Mutation | ACC,GCC | T,A 654 | XP_016869657.1 |