Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCCTCTGGCAAAACTCCCAGA[C/T]GGTGGCGTTGACCGTGCTCAGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616748 MIM: 611255 | ||||||||||||||||||||
Literature Links: |
ENTPD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENTPD8 - ectonucleoside triphosphate diphosphohydrolase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033113.1 | 974 | Missense Mutation | ATC,GTC | I,V 377 | NP_001028285.1 | |
NM_198585.2 | 974 | Intron | NP_940987.2 | |||
XM_011518665.2 | 974 | Missense Mutation | ATC,GTC | I,V 485 | XP_011516967.1 | |
XM_011518666.2 | 974 | Intron | XP_011516968.1 | |||
XM_011518668.1 | 974 | Missense Mutation | ATC,GTC | I,V 250 | XP_011516970.1 |
NOXA1 - NADPH oxidase activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |