Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCTGGGCTCACCAGGCCGGGCC[A/G]CCAGGTAGAGACCAGTGTCTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609379 MIM: 612902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCN15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCN15 - lipocalin 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCN6 - lipocalin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCN8 - lipocalin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178469.3 | 759 | Missense Mutation | GCG,GTG | A,V 137 | NP_848564.2 | |
XM_017014272.1 | 759 | Missense Mutation | GCG,GTG | A,V 160 | XP_016869761.1 |
LOC100128593 - uncharacterized LOC100128593 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6722 - microRNA 6722 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |