Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCCTTTACCTTTGAAAGGAAG[A/G]ATCCTGTAAGTACAATCCCAAGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116880 | ||||||||||||||||||||
Literature Links: |
CTSL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTSL - cathepsin L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257971.1 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | NP_001244900.1 | |
NM_001257972.1 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | NP_001244901.1 | |
NM_001257973.1 | 1516 | Missense Mutation | GAA,GGA | E,G 27 | NP_001244902.1 | |
NM_001912.4 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | NP_001903.1 | |
NM_145918.2 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | NP_666023.1 | |
XM_005251716.3 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | XP_005251773.1 | |
XM_011518263.1 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | XP_011516565.1 | |
XM_017014293.1 | 1516 | Missense Mutation | GAA,GGA | E,G 209 | XP_016869782.1 |