Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCCTCGATGGCGATGCCCAGG[C/G]TGGCCGCACTTTTCTTCACACGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 607928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WHRN - whirlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083885.2 | 3215 | Missense Mutation | ACC,AGC | T,S 442 | NP_001077354.2 | |
NM_001173425.1 | 3215 | Missense Mutation | ACC,AGC | T,S 824 | NP_001166896.1 | |
NM_015404.3 | 3215 | Missense Mutation | ACC,AGC | T,S 825 | NP_056219.3 | |
XM_011518484.1 | 3215 | Missense Mutation | ACC,AGC | T,S 836 | XP_011516786.1 | |
XM_011518485.1 | 3215 | Missense Mutation | ACC,AGC | T,S 836 | XP_011516787.1 | |
XM_011518486.1 | 3215 | Missense Mutation | ACC,AGC | T,S 835 | XP_011516788.1 | |
XM_011518487.1 | 3215 | Missense Mutation | ACC,AGC | T,S 794 | XP_011516789.1 | |
XM_011518488.1 | 3215 | Missense Mutation | ACC,AGC | T,S 755 | XP_011516790.1 | |
XM_011518489.2 | 3215 | Intron | XP_011516791.1 | |||
XM_011518491.2 | 3215 | Intron | XP_011516793.1 | |||
XM_011518492.1 | 3215 | Intron | XP_011516794.1 | |||
XM_011518494.2 | 3215 | Intron | XP_011516796.1 | |||
XM_011518495.1 | 3215 | Missense Mutation | ACC,AGC | T,S 395 | XP_011516797.1 |