Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGCAAGTGAAAGTTCTTCTAA[C/T]AGAGAAAGTTTTGAAAAAGTACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 MIM: 602383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | 783 | Intron | NP_001012267.1 | |||
NM_001286969.1 | 783 | Intron | NP_001273898.1 | |||
NM_001286971.1 | 783 | Intron | NP_001273900.1 | |||
XM_011518685.2 | 783 | Intron | XP_011516987.1 | |||
XM_011518689.1 | 783 | Intron | XP_011516991.1 | |||
XM_017014715.1 | 783 | Intron | XP_016870204.1 |
OGN - osteoglycin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014057.4 | 783 | Silent Mutation | CTA,CTG | L,L 168 | NP_054776.1 | |
NM_024416.4 | 783 | Silent Mutation | CTA,CTG | L,L 226 | NP_077727.3 | |
NM_033014.3 | 783 | Silent Mutation | CTA,CTG | L,L 168 | NP_148935.1 |