Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCACTGTCTGGGTTCAGGTCCAC[A/G]AACATCTTCAGAATGAATTCTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNAQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNAQ - G protein subunit alpha q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002072.4 | 1153 | Silent Mutation | TTC,TTT | F,F 313 | NP_002063.2 | |
XM_017014628.1 | 1153 | Silent Mutation | TTC,TTT | F,F 255 | XP_016870117.1 |