Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGCCTGCGCGCTCCAGGTCCT[C/G]CGGCAGGATGCGGCCCTTCTTGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185641 MIM: 185642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED22 - mediator complex subunit 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF6 - surfeit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278942.1 | 1135 | UTR 3 | NP_001265871.1 | |||
NM_006753.5 | 1135 | Missense Mutation | CAG,GAG | Q,E 353 | NP_006744.2 |