Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGAAGGATGGCCATTTTCAA[A/C]TGAGGACGTAGCTCCATTCATCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604296 MIM: 616590 | ||||||||||||||||||||
Literature Links: |
GRHPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRHPR - glyoxylate reductase/hydroxypyruvate reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB5 - zinc finger and BTB domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014872.2 | 1911 | Missense Mutation | GTT,TTT | V,F 571 | NP_055687.1 | |
XM_005251634.1 | 1911 | Missense Mutation | GTT,TTT | V,F 571 | XP_005251691.1 |