Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGGTTAATAACTGTGAGAACAC[A/G]GGCAATGGATTTCCGGACGACTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ARPC5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARPC5L - actin related protein 2/3 complex subunit 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL35 - ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007209.3 | Intron | NP_009140.1 |
WDR38 - WD repeat domain 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |