Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCCTGCAGGAGGAACCGGTGGA[A/G]GGCTTCCGGATCACCCTGGTGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2R2 - ubiquitin conjugating enzyme E2 R2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017811.3 | 672 | Silent Mutation | GAA,GAG | E,E 26 | NP_060281.2 | |
XM_005251496.2 | 672 | Silent Mutation | GAA,GAG | E,E 26 | XP_005251553.1 | |
XM_011517949.1 | 672 | Silent Mutation | GAA,GAG | E,E 26 | XP_011516251.1 | |
XM_017014860.1 | 672 | Silent Mutation | GAA,GAG | E,E 26 | XP_016870349.1 |