Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCACTCACCTGCACACCTGCCT[A/G]GCAAGGGCAGGGAAGAGCCGCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190198 MIM: 612854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf163 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf163 - chromosome 9 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152571.2 | 73 | Nonsense Mutation | TAG,TGG | *,W 11 | NP_689784.1 |
NOTCH1 - notch 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |