Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTTCTGCCCATTGTGGGCTGC[C/T]GAGGGACGTCTGAGAGAATTTTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO10 - F-box protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012166.2 | 3132 | Silent Mutation | TCA,TCG | S,S 922 | NP_036298.2 | |
XM_005251439.4 | 3132 | Intron | XP_005251496.1 | |||
XM_006716754.3 | 3132 | Silent Mutation | TCA,TCG | S,S 447 | XP_006716817.1 | |
XM_017014618.1 | 3132 | Silent Mutation | TCA,TCG | S,S 980 | XP_016870107.1 | |
XM_017014619.1 | 3132 | Silent Mutation | TCA,TCG | S,S 922 | XP_016870108.1 |
POLR1E - RNA polymerase I subunit E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |