Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATGATGCTGATGATTATCACCC[A/G]GAGGAAAATCGAAGGAAGATGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611343 | ||||||||||||||||||||
Literature Links: |
IDNK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDNK - IDNK, gluconokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001551.3 | 937 | Silent Mutation | CCA,CCG | P,P 40 | NP_001001551.2 | |
NM_001256915.1 | 937 | UTR 5 | NP_001243844.1 | |||
XM_006717110.3 | 937 | Silent Mutation | CCA,CCG | P,P 83 | XP_006717173.1 | |
XM_006717111.3 | 937 | Silent Mutation | CCA,CCG | P,P 41 | XP_006717174.1 | |
XM_017014719.1 | 937 | Intron | XP_016870208.1 | |||
XM_017014720.1 | 937 | Silent Mutation | CCA,CCG | P,P 80 | XP_016870209.1 | |
XM_017014721.1 | 937 | Silent Mutation | CCA,CCG | P,P 164 | XP_016870210.1 | |
XM_017014722.1 | 937 | Intron | XP_016870211.1 | |||
XM_017014723.1 | 937 | Intron | XP_016870212.1 | |||
XM_017014724.1 | 937 | UTR 5 | XP_016870213.1 | |||
XM_017014725.1 | 937 | UTR 5 | XP_016870214.1 |