Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCGAGTCTGGGGCCGGGGCCA[A/G]GGCCGGACTCGGGCTCACGTCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609720 MIM: 613633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRB2 - crumbs 2, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DENND1A - DENN domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020946.1 | 4081 | Silent Mutation | CTG,TTG | L,L 991 | NP_065997.1 | |
NM_024820.2 | 4081 | Intron | NP_079096.2 | |||
XM_005252109.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1052 | XP_005252166.1 | |
XM_005252111.4 | 4081 | Silent Mutation | CTG,TTG | L,L 1006 | XP_005252168.1 | |
XM_005252113.4 | 4081 | Intron | XP_005252170.1 | |||
XM_006717195.3 | 4081 | Silent Mutation | CTG,TTG | L,L 1049 | XP_006717258.1 | |
XM_011518882.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1067 | XP_011517184.1 | |
XM_011518883.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1048 | XP_011517185.1 | |
XM_011518884.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1038 | XP_011517186.1 | |
XM_011518885.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1031 | XP_011517187.1 | |
XM_011518886.2 | 4081 | Silent Mutation | CTG,TTG | L,L 1024 | XP_011517188.1 | |
XM_011518887.2 | 4081 | Intron | XP_011517189.1 | |||
XM_017014947.1 | 4081 | Silent Mutation | CTG,TTG | L,L 1040 | XP_016870436.1 | |
XM_017014948.1 | 4081 | Silent Mutation | CTG,TTG | L,L 1034 | XP_016870437.1 | |
XM_017014949.1 | 4081 | Silent Mutation | CTG,TTG | L,L 1025 | XP_016870438.1 | |
XM_017014950.1 | 4081 | Silent Mutation | CTG,TTG | L,L 1007 | XP_016870439.1 | |
XM_017014951.1 | 4081 | Silent Mutation | CTG,TTG | L,L 1002 | XP_016870440.1 | |
XM_017014952.1 | 4081 | Silent Mutation | CTG,TTG | L,L 793 | XP_016870441.1 |