Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGAGTGGTGTCAGACCTAATG[A/G]CAGTCCTGTCTGACTGCATAACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL13 - kelch like family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168299.1 | 1803 | Missense Mutation | NP_001161771.1 | |||
NM_001168300.1 | 1803 | Missense Mutation | NP_001161772.1 | |||
NM_001168301.1 | 1803 | Missense Mutation | NP_001161773.1 | |||
NM_001168302.1 | 1803 | Missense Mutation | NP_001161774.1 | |||
NM_001168303.1 | 1803 | Missense Mutation | NP_001161775.1 | |||
NM_033495.3 | 1803 | Missense Mutation | NP_277030.2 | |||
XM_011531409.2 | 1803 | Missense Mutation | XP_011529711.1 | |||
XM_011531410.2 | 1803 | Missense Mutation | XP_011529712.1 | |||
XM_011531411.1 | 1803 | Missense Mutation | XP_011529713.1 | |||
XM_017029950.1 | 1803 | Missense Mutation | XP_016885439.1 |