Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGATGCCGAGTCTTCTCCAGA[G/T]TTCCTCTGAGAGTCCTCAGAGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300223 MIM: 300469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEC1 - MAGE family member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005462.4 | 374 | Missense Mutation | AGT,ATT | S,I 17 | NP_005453.2 | |
XM_011531418.2 | 374 | Missense Mutation | AGT,ATT | S,I 17 | XP_011529720.1 |
MAGEC3 - MAGE family member C3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |