Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATAATGGAGTCTGAGTATGTCC[A/T]ATGCAACTGGAAAGACCAGTTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MUM1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUM1L1 - MUM1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171020.1 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | NP_001164491.1 | |
NM_152423.4 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | NP_689636.3 | |
XM_005262077.2 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | XP_005262134.1 | |
XM_011530856.1 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | XP_011529158.1 | |
XM_011530857.1 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | XP_011529159.1 | |
XM_017029269.1 | 663 | Missense Mutation | CAA,CTA | Q,L 7 | XP_016884758.1 |