Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAATCGGCCAGGCTGTCGTCACT[A/G]CCCAGGGGCTTGATGTCCCCGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 308840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
L1CAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
L1CAM - L1 cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000425.4 | 3827 | Silent Mutation | GGC,GGT | G,G 1203 | NP_000416.1 | |
NM_001143963.2 | 3827 | Silent Mutation | GGC,GGT | G,G 1194 | NP_001137435.1 | |
NM_001278116.1 | 3827 | Silent Mutation | GGC,GGT | G,G 1203 | NP_001265045.1 | |
NM_024003.3 | 3827 | Silent Mutation | GGC,GGT | G,G 1199 | NP_076493.1 |