Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACGAAATGCAAGATCATTACTA[C/T]CGTAAGTACCTGTGTTTTGCCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOO - apolipoprotein O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CXorf58 - chromosome X open reading frame 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169574.1 | 216 | Missense Mutation | CCA,TCA | P,S 39 | NP_001163045.1 | |
NM_152761.2 | 216 | Missense Mutation | CCA,TCA | P,S 39 | NP_689974.2 | |
XM_011545473.2 | 216 | Missense Mutation | CCA,TCA | P,S 39 | XP_011543775.1 | |
XM_011545474.2 | 216 | Intron | XP_011543776.1 |