Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTTGGCCTCCCTGGTCCGTGG[C/G]TGTCTCCACGCCTTCCGGGGGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP6 - Rho GTPase activating protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287242.1 | 2369 | Missense Mutation | CCC,GCC | P,A 727 | NP_001274171.1 | |
NM_006125.2 | 2369 | Intron | NP_006116.2 | |||
NM_013423.2 | 2369 | Missense Mutation | CCC,GCC | P,A 704 | NP_038267.1 | |
NM_013427.2 | 2369 | Missense Mutation | CCC,GCC | P,A 907 | NP_038286.2 |