Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGGCTGGGACCAAGCAGCGTC[A/G]TCTGTGTCCAGAGGGTCCCATGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300023 MIM: 300538 | ||||||||||||||||||||
Literature Links: |
ARHGAP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP4 - Rho GTPase activating protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164741.1 | 2735 | Nonsense Mutation | CGA,TGA | R,* 893 | NP_001158213.1 | |
NM_001666.4 | 2735 | Nonsense Mutation | CGA,TGA | R,* 853 | NP_001657.3 |
AVPR2 - arginine vasopressin receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |