Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGACCGCCGCCCCCTCCGCCGC[C/G]TCCCCATCCCCACGCTCGCATCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 313700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AR - androgen receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000044.3 | 2254 | Missense Mutation | CCT,CGT | P,R 380 | NP_000035.2 | |
NM_001011645.2 | 2254 | Intron | NP_001011645.1 |