Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACTGAATGAGGTATCATCAGGC[A/T]CCAGTGAGGCATGATTGAAATCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HDX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HDX - highly divergent homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177478.1 | 2109 | Missense Mutation | GAG,GTG | E,V 612 | NP_001170949.1 | |
NM_001177479.1 | 2109 | Missense Mutation | GAG,GTG | E,V 670 | NP_001170950.1 | |
NM_144657.4 | 2109 | Missense Mutation | GAG,GTG | E,V 670 | NP_653258.2 | |
XM_006724619.1 | 2109 | Missense Mutation | GAG,GTG | E,V 612 | XP_006724682.1 | |
XM_011530863.2 | 2109 | Missense Mutation | GAG,GTG | E,V 670 | XP_011529165.1 | |
XM_011530864.1 | 2109 | Missense Mutation | GAG,GTG | E,V 670 | XP_011529166.1 | |
XM_017029274.1 | 2109 | Missense Mutation | GAG,GTG | E,V 670 | XP_016884763.1 | |
XM_017029275.1 | 2109 | Missense Mutation | GAG,GTG | E,V 652 | XP_016884764.1 |