Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAAAACAGGACTTCACAAGGTA[A/G]TAGTAGTTTTCAGACCACGCTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300065 | ||||||||||||||||||||
Literature Links: |
CENPI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPI - centromere protein I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318521.1 | 196 | Missense Mutation | AAT,AGT | N,S 20 | NP_001305450.1 | |
NM_001318523.1 | 196 | Missense Mutation | AAT,AGT | N,S 20 | NP_001305452.1 | |
NM_006733.3 | 196 | Missense Mutation | AAT,AGT | N,S 20 | NP_006724.2 | |
XM_005262111.2 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_005262168.1 | |
XM_011530894.2 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_011529196.1 | |
XM_011530895.2 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_011529197.1 | |
XM_011530897.2 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_011529199.1 | |
XM_011530899.2 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_011529201.1 | |
XM_017029383.1 | 196 | Missense Mutation | AAT,AGT | N,S 20 | XP_016884872.1 |
TMEM35A - transmembrane protein 35A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |