Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGTAGGAGAAGCTACCTGTAG[C/T]GCTAGAACTTGCACTGGCCATGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300343 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEA10 - MAGE family member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011543.2 | 1514 | Missense Mutation | ACT,GCT | T,A 361 | NP_001011543.2 | |
NM_001251828.1 | 1514 | Missense Mutation | ACT,GCT | T,A 361 | NP_001238757.1 | |
NM_021048.4 | 1514 | Missense Mutation | ACT,GCT | T,A 361 | NP_066386.2 |
MAGEA10-MAGEA5 - MAGEA10-MAGEA5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204811.2 | 1514 | Intron | NP_001191740.1 |