Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTTCTCTGGGAAATTCTGCAGG[C/G]CTTGTAGTTCTCTAAGCCCCTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311040 MIM: 300234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UXT - ubiquitously expressed prefoldin like chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004182.3 | 693 | Missense Mutation | GCC,GGC | A,G 147 | NP_004173.1 | |
NM_153477.2 | 693 | Missense Mutation | GCC,GGC | A,G 159 | NP_705582.1 |
UXT-AS1 - UXT antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |