Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTATCTTTCTGAAACAAGTTT[C/T]TGGCTCTAGTCCTCTGGCTTTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZMAT1 - zinc finger matrin-type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011657.3 | 1598 | Missense Mutation | AAA,GAA | K,E 411 | NP_001011657.2 | |
NM_001282400.1 | 1598 | Missense Mutation | AAA,GAA | K,E 240 | NP_001269329.1 | |
NM_001282401.1 | 1598 | Missense Mutation | AAA,GAA | K,E 240 | NP_001269330.1 | |
XM_005262212.4 | 1598 | Missense Mutation | AAA,GAA | K,E 491 | XP_005262269.2 | |
XM_005262213.4 | 1598 | Missense Mutation | AAA,GAA | K,E 490 | XP_005262270.2 | |
XM_005262214.3 | 1598 | Missense Mutation | AAA,GAA | K,E 468 | XP_005262271.2 | |
XM_005262216.4 | 1598 | Missense Mutation | AAA,GAA | K,E 371 | XP_005262273.2 | |
XM_006724711.3 | 1598 | Missense Mutation | AAA,GAA | K,E 240 | XP_006724774.1 | |
XM_017029903.1 | 1598 | Missense Mutation | AAA,GAA | K,E 370 | XP_016885392.1 | |
XM_017029904.1 | 1598 | Missense Mutation | AAA,GAA | K,E 240 | XP_016885393.1 |