Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACGTGTTGTTGACTCTTCGACCA[C/T]GGCGTATTAATATGACAGACACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300359 | ||||||||||||||||||||
Literature Links: |
SAGE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAGE1 - sarcoma antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018666.2 | 857 | Missense Mutation | CGG,TGG | R,W 228 | NP_061136.2 | |
XM_017029621.1 | 857 | Missense Mutation | CGG,TGG | R,W 228 | XP_016885110.1 | |
XM_017029622.1 | 857 | Intron | XP_016885111.1 | |||
XM_017029623.1 | 857 | Missense Mutation | CGG,TGG | R,W 228 | XP_016885112.1 | |
XM_017029624.1 | 857 | Missense Mutation | CGG,TGG | R,W 197 | XP_016885113.1 | |
XM_017029625.1 | 857 | Missense Mutation | CGG,TGG | R,W 228 | XP_016885114.1 | |
XM_017029626.1 | 857 | Intron | XP_016885115.1 | |||
XM_017029627.1 | 857 | Intron | XP_016885116.1 |