Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCATGAGTCGTATGTAAAGTA[C/G]TACATGGCAAATCCAACAGTCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOOL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOOL - apolipoprotein O like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198450.5 | 1803 | Intron | NP_940852.3 | |||
XM_011530859.2 | 1803 | Intron | XP_011529161.1 | |||
XM_017029271.1 | 1803 | Intron | XP_016884760.1 | |||
XM_017029272.1 | 1803 | Intron | XP_016884761.1 | |||
XM_017029273.1 | 1803 | Intron | XP_016884762.1 |
SATL1 - spermidine/spermine N1-acetyl transferase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012980.2 | 1803 | Nonsense Mutation | TAC,TAG | Y,* 601 | NP_001012998.2 | |
XM_011530940.2 | 1803 | Nonsense Mutation | TAC,TAG | Y,* 601 | XP_011529242.1 |