Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGGCTATGAATATTCACTTTC[C/T]GTTTTCTTTCCAGAAAGTGGTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300170 MIM: 300202 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OFD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003611.2 | 566 | Silent Mutation | TCC,TCT | S,S 92 | NP_003602.1 | |
XM_005274604.2 | 566 | Silent Mutation | TCC,TCT | S,S 92 | XP_005274661.1 | |
XM_005274606.3 | 566 | Silent Mutation | TCC,TCT | S,S 44 | XP_005274663.1 | |
XM_011545592.2 | 566 | Intron | XP_011543894.1 | |||
XM_011545594.2 | 566 | Intron | XP_011543896.1 | |||
XM_011545595.1 | 566 | Intron | XP_011543897.1 | |||
XM_011545597.1 | 566 | UTR 5 | XP_011543899.1 | |||
XM_011545598.1 | 566 | Intron | XP_011543900.1 | |||
XM_017029909.1 | 566 | Intron | XP_016885398.1 | |||
XM_017029910.1 | 566 | Intron | XP_016885399.1 | |||
XM_017029911.1 | 566 | Intron | XP_016885400.1 |
TRAPPC2 - trafficking protein particle complex 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |