Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGACAGACCATTGCATGAAATCG[A/G]AACTGACTACACTTTTGTTAGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ARL13A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL13A - ADP ribosylation factor like GTPase 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162491.1 | 979 | Missense Mutation | AAA,GAA | K,E 56 | NP_001155963.1 | |
XM_011530950.2 | 979 | Missense Mutation | AAA,GAA | K,E 56 | XP_011529252.1 | |
XM_011530951.2 | 979 | Missense Mutation | AAA,GAA | K,E 56 | XP_011529253.1 |