Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAGACCTTTACGAATTGGTCAC[A/G]TAGCAGCTTCATCTCTTTCAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM9B - family with sequence similarity 9 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205849.3 | 1413 | Missense Mutation | CGT,TGT | R,C 159 | NP_995321.1 | |
XM_011545464.2 | 1413 | Missense Mutation | CGT,TGT | R,C 159 | XP_011543766.1 | |
XM_011545465.2 | 1413 | Intron | XP_011543767.1 |