Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTGCGATGCTGCTGGGCGACC[A/G]GATCAGGTAGGCTGCCCCCGCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300050 | ||||||||||||||||||||
Literature Links: |
USP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP11 - ubiquitin specific peptidase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004651.3 | 1302 | Silent Mutation | CCA,CCG | P,P 434 | NP_004642.2 | |
XM_005272674.3 | 1302 | Silent Mutation | CCA,CCG | P,P 161 | XP_005272731.1 | |
XM_011543988.1 | 1302 | Silent Mutation | CCA,CCG | P,P 161 | XP_011542290.1 |