Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATGGCCAAACTTGCTGAACATA[A/T]GTTCCCAAAGAGCCAGGAATAACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300364 | ||||||||||||||||||||
Literature Links: |
ARMCX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMCX3 - armadillo repeat containing, X-linked 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016607.3 | 1780 | Missense Mutation | AAG,ATG | K,M 373 | NP_057691.1 | |
NM_177947.2 | 1780 | Missense Mutation | AAG,ATG | K,M 373 | NP_808816.1 | |
NM_177948.2 | 1780 | Missense Mutation | AAG,ATG | K,M 373 | NP_808817.1 | |
XM_005262141.2 | 1780 | Missense Mutation | AAG,ATG | K,M 373 | XP_005262198.1 |
ARMCX6 - armadillo repeat containing, X-linked 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |