Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGACCGTGATGACGGACGCTACC[C/T]GCCCAGGCCCAGGCCACGGCCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XG - Xg blood group | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141919.1 | 1447 | Intron | NP_001135391.1 | |||
NM_001141920.1 | 1447 | Intron | NP_001135392.1 | |||
NM_175569.2 | 1447 | Intron | NP_780778.1 | |||
XM_005274587.3 | 1447 | Missense Mutation | CCG,CTG | P,L 99 | XP_005274644.1 | |
XM_011545575.2 | 1447 | Missense Mutation | CCG,CTG | P,L 99 | XP_011543877.1 | |
XM_017029787.1 | 1447 | Missense Mutation | CCG,CTG | P,L 85 | XP_016885276.1 |