Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGAAGGGCACACATCTTAGGACC[C/T]GGAGGACGTCCGGCCTCTGTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A3 - solute carrier family 25 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002635.3 | 47 | UTR 5 | NP_002626.1 | |||
NM_005888.3 | 47 | UTR 5 | NP_005879.1 | |||
NM_213611.2 | 47 | UTR 5 | NP_998776.1 |
SNORA53 - small nucleolar RNA, H/ACA box 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |