Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGGGGGTCTCCTCCAGGGGTC[G/A]CGGGGCCCGGGGCCGCCCCAGCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605217 | ||||||||||||||||||||
Literature Links: |
ODF3L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ODF3L2 - outer dense fiber of sperm tails 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182577.2 | 500 | Nonsense Mutation | CGA,TGA | R,* 232 | NP_872383.1 | |
XM_005259545.4 | 500 | Nonsense Mutation | CGA,TGA | R,* 124 | XP_005259602.1 | |
XM_017026671.1 | 500 | Nonsense Mutation | CGA,TGA | R,* 124 | XP_016882160.1 |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |