Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGCTTTGTGGTTCATCGACGA[T/A]GCCACGAATTTGTGACCTTCGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609959 MIM: 176980 | ||||||||||||||||||||
Literature Links: |
MYADM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYADM - myeloid associated differentiation marker | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKCG - protein kinase C gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316329.1 | 540 | Missense Mutation | AGC,TGC | S,C 77 | NP_001303258.1 | |
NM_002739.4 | 540 | Missense Mutation | AGC,TGC | S,C 77 | NP_002730.1 |