Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAGAATAGATTTTAAAAAGAAA[G/T]TAATAATAACTTGACTTGGAAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBM24 - RNA binding motif protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143941.1 | Intron | NP_001137413.1 | ||||
NM_001143942.1 | Intron | NP_001137414.1 | ||||
NM_153020.2 | Intron | NP_694565.1 | ||||
XM_005248932.3 | Intron | XP_005248989.1 | ||||
XM_011514387.2 | Intron | XP_011512689.1 | ||||
XM_011514388.2 | Intron | XP_011512690.1 | ||||
XM_011514389.2 | Intron | XP_011512691.1 |