Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614919 MIM: 180661 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOA1 - nitric oxide associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2B - RNA polymerase II subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000938.2 | 97 | UTR 5 | NP_000929.1 | |||
NM_001303268.1 | 97 | UTR 5 | NP_001290197.1 | |||
NM_001303269.1 | 97 | UTR 5 | NP_001290198.1 |
Set Membership: |
HapMap |