Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAAGTGACCCTTGAGAAGTTTA[C/T]TTTTTGGTTAGTGACCTATTGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605774 MIM: 607545 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL2 - kelch like family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSMO1 - methylsterol monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017369.2 | Intron | NP_001017369.1 | ||||
NM_006745.4 | Intron | NP_006736.1 | ||||
XM_005263176.2 | Intron | XP_005263233.1 |
Set Membership: |
HapMap |