Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAAGGTCTGTGATTGTATGTGA[C/T]CTTAGTCATCTATTTGTTACACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616919 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMPD1 - FERM and PDZ domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014907.2 | Intron | NP_055722.2 | ||||
XM_011517803.2 | Intron | XP_011516105.1 | ||||
XM_011517804.2 | Intron | XP_011516106.1 | ||||
XM_011517805.2 | Intron | XP_011516107.1 | ||||
XM_011517806.2 | Intron | XP_011516108.1 | ||||
XM_017014480.1 | Intron | XP_016869969.1 | ||||
XM_017014481.1 | Intron | XP_016869970.1 | ||||
XM_017014482.1 | Intron | XP_016869971.1 |