Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602559 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
XPO1 - exportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003400.3 | Intron | NP_003391.1 | ||||
XM_005264544.1 | Intron | XP_005264601.1 | ||||
XM_005264546.1 | Intron | XP_005264603.1 | ||||
XM_006712094.2 | Intron | XP_006712157.1 | ||||
XM_011533097.1 | Intron | XP_011531399.1 | ||||
XM_011533098.1 | Intron | XP_011531400.1 | ||||
XM_011533099.2 | Intron | XP_011531401.1 |
Set Membership: |
HapMap |