Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613639 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADGRD1 - adhesion G protein-coupled receptor D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253566.2 | Intron | XP_005253623.1 | ||||
XM_011538203.2 | Intron | XP_011536505.1 | ||||
XM_011538204.2 | Intron | XP_011536506.1 | ||||
XM_011538205.2 | Intron | XP_011536507.1 | ||||
XM_011538206.2 | Intron | XP_011536508.1 | ||||
XM_011538207.2 | Intron | XP_011536509.1 | ||||
XM_011538208.2 | Intron | XP_011536510.1 | ||||
XM_011538209.2 | Intron | XP_011536511.1 | ||||
XM_011538210.2 | Intron | XP_011536512.1 | ||||
XM_011538211.2 | Intron | XP_011536513.1 | ||||
XM_011538212.2 | Intron | XP_011536514.1 |
Set Membership: |
HapMap Validated |