Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCTCCAGAGGTTGAGGTCGGGC[C/T]CATCTCCAGTTTCACCCAGGGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRB2 - crumbs 2, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173689.6 | Intron | NP_775960.4 | ||||
XM_005251934.2 | Intron | XP_005251991.1 | ||||
XM_011518556.2 | Intron | XP_011516858.1 | ||||
XM_011518557.2 | Intron | XP_011516859.1 | ||||
XM_011518558.2 | Intron | XP_011516860.1 |