Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612120 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIDEC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CIDEC - cell death inducing DFFA like effector c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199551.1 | 872 | UTR 3 | NP_001186480.1 | |||
NM_001199552.1 | 872 | UTR 3 | NP_001186481.1 | |||
NM_001199623.1 | 872 | UTR 3 | NP_001186552.1 | |||
NM_001321142.1 | 872 | UTR 3 | NP_001308071.1 | |||
NM_001321143.1 | 872 | UTR 3 | NP_001308072.1 | |||
NM_001321144.1 | 872 | UTR 3 | NP_001308073.1 | |||
NM_022094.3 | 872 | UTR 3 | NP_071377.2 |