Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCAGCAACTTGAAGAGGTGGTC[A/G]TAGGCTTTGGCCATGGCGGACACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116953 MIM: 179514 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK2 - cyclin dependent kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252036.1 | Intron | NP_001238965.1 | ||||
NM_001252037.1 | Intron | NP_001238966.1 | ||||
NM_002868.3 | Intron | NP_002859.1 | ||||
XM_005269051.2 | Intron | XP_005269108.1 | ||||
XM_005269052.2 | Intron | XP_005269109.1 |